Product Details

SNP ID
rs4905374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95687468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCTCTACTATATTTGAAATCAG[A/G]TTACTAATAACACCTAATGCTACAC
Phenotype
MIM: 186960 MIM: 603769
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCL1A PubMed Links

Gene Details

Gene
TCL1A
Gene Name
T-cell leukemia/lymphoma 1A
There are no transcripts associated with this gene.

Gene
TCL1B
Gene Name
T-cell leukemia/lymphoma 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004918.3 Intron NP_004909.1

View Full Product Details