Product Details

SNP ID
rs8077936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75209150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTTTTATCCCTTTTTCTTTGTCC[A/G]AAGTGACCAAATTCCTATCAGTAGC
Phenotype
MIM: 170285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP85 PubMed Links

Gene Details

Gene
NUP85
Gene Name
nucleoporin 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303276.1 Intron NP_001290205.1
NM_024844.4 Intron NP_079120.1
XM_005257690.2 Intron XP_005257747.1
XM_005257692.3 Intron XP_005257749.1
XM_006722094.1 Intron XP_006722157.1
XM_011525267.1 Intron XP_011523569.1
XM_011525268.1 Intron XP_011523570.1
XM_017025108.1 Intron XP_016880597.1
XM_017025109.1 Intron XP_016880598.1
XM_017025110.1 Intron XP_016880599.1

View Full Product Details