Product Details

SNP ID
rs3776801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115523956 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGCAAGAAAAAAAGACTTTCAA[C/T]TGTATAAAATTCACAAACCAGTAAA
Phenotype
MIM: 608767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FEM1C PubMed Links

Gene Details

Gene
FEM1C
Gene Name
fem-1 homolog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020177.2 2854 UTR 3 NP_064562.1
XM_005272035.4 2854 UTR 3 XP_005272092.1
XM_017009647.1 2854 UTR 3 XP_016865136.1

View Full Product Details