Product Details

SNP ID
rs4268692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99693004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACCTTGGACAAGTTGCTTCATT[C/T]TGGGCCTCCATTTCCTCATCTGTGA
Phenotype
MIM: 604087
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP46A1 PubMed Links

Gene Details

Gene
CYP46A1
Gene Name
cytochrome P450 family 46 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006668.1 Intron NP_006659.1

View Full Product Details