Product Details

SNP ID
rs7938320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:44104780 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAATGGCAAGGAAGCCATACCA[C/T]AGGGTGGGCCTGCAGCTGGTAGGGG
Phenotype
MIM: 608210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXT2 PubMed Links

Gene Details

Gene
EXT2
Gene Name
exostosin glycosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000401.3 Intron NP_000392.3
NM_001178083.1 Intron NP_001171554.1
NM_207122.1 Intron NP_997005.1
XM_011519950.1 Intron XP_011518252.1
XM_011519951.1 Intron XP_011518253.1

View Full Product Details