Product Details

SNP ID
rs3218701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108310089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAGTCACTACCATTGTATTCT[A/G]TATCAACATGCTTTTATTTTGATAT
Phenotype
MIM: 607585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATM PubMed Links

Gene Details

Gene
ATM
Gene Name
ATM serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000051.3 Intron NP_000042.3
XM_005271561.4 Intron XP_005271618.2
XM_005271562.4 Intron XP_005271619.2
XM_006718843.3 Intron XP_006718906.1
XM_006718845.1 Intron XP_006718908.1
XM_011542840.2 Intron XP_011541142.1
XM_011542842.2 Intron XP_011541144.1
XM_011542843.2 Intron XP_011541145.1
XM_011542844.2 Intron XP_011541146.1
XM_011542845.1 Intron XP_011541147.1
XM_017017789.1 Intron XP_016873278.1
XM_017017790.1 Intron XP_016873279.1
XM_017017791.1 Intron XP_016873280.1
XM_017017792.1 Intron XP_016873281.1
Gene
C11orf65
Gene Name
chromosome 11 open reading frame 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152587.3 Intron NP_689800.3
XM_005271412.3 Intron XP_005271469.1
XM_005271413.3 Intron XP_005271470.1
XM_005271414.3 Intron XP_005271471.1
XM_005271415.4 Intron XP_005271472.1
XM_011542639.2 Intron XP_011540941.1
XM_011542640.2 Intron XP_011540942.1
XM_011542641.2 Intron XP_011540943.1
XM_011542642.1 Intron XP_011540944.1
XM_011542643.2 Intron XP_011540945.1
XM_017017246.1 Intron XP_016872735.1
XM_017017247.1 Intron XP_016872736.1
XM_017017248.1 Intron XP_016872737.1
XM_017017249.1 Intron XP_016872738.1
XM_017017250.1 Intron XP_016872739.1

View Full Product Details