Product Details

SNP ID
rs3213038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:60782942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGCCACTGGTTAATTGTTAATTC[C/T]GAAAATTATTTAGGATTAGATGAGA
Phenotype
MIM: 116940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK1 PubMed Links

Gene Details

Gene
CDK1
Gene Name
cyclin dependent kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170406.1 Intron NP_001163877.1
NM_001170407.1 Intron NP_001163878.1
NM_001320918.1 Intron NP_001307847.1
NM_001786.4 Intron NP_001777.1
NM_033379.4 Intron NP_203698.1
XM_005270303.3 Intron XP_005270360.1

View Full Product Details