Product Details

SNP ID
rs4142048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68284274 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAATTCCCAGCTCTCCTCCTCGG[T/C]GGGAACACTGAAAAGCTAAAGAAAA
Phenotype
MIM: 612189
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
PBLD PubMed Links

Gene Details

Gene
PBLD
Gene Name
phenazine biosynthesis like protein domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033083.1 941 Intron NP_001028255.1
NM_022129.3 941 Missense Mutation CAC,CGC H257R NP_071412.2
XM_005270028.3 941 Missense Mutation CAC,CGC H257R XP_005270085.1
XM_011540060.2 941 Missense Mutation ACC,GCC T249A XP_011538362.1
XM_017016513.1 941 Missense Mutation ACC,GCC T249A XP_016872002.1
XM_017016514.1 941 Intron XP_016872003.1

View Full Product Details