Product Details

SNP ID
rs3917575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95298684 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTTAATATCTGACAATTGACAT[A/G]ACTGATTTATCCATTTTAGTGAGAA
Phenotype
MIM: 168820 MIM: 602468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PON1 PubMed Links

Gene Details

Gene
PON1
Gene Name
paraoxonase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000446.5 1425 UTR 3 NP_000437.3
Gene
PPP1R9A
Gene Name
protein phosphatase 1 regulatory subunit 9A
There are no transcripts associated with this gene.

View Full Product Details