Product Details

SNP ID
rs3917635
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:94529533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAAAAATATAGTTAGCTCTCAAA[C/T]GTTTAATGCCACTTAAGTCAGTTAA
Phenotype
MIM: 134390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F3 PubMed Links

Gene Details

Gene
F3
Gene Name
coagulation factor III, tissue factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178096.1 2036 UTR 3 NP_001171567.1
NM_001993.4 2036 UTR 3 NP_001984.1

View Full Product Details