Product Details

SNP ID
hCV32340905
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.1:229518281 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TTCCATATTCAGTAATTTTTCCTTG[A/G]TCAAGAACAGCAACCATATTAGCAT
Phenotype
MIM: 605454 MIM: 607613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCB10 PubMed Links
Additional Information
For this assay, SNP(s) [rs115488729] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ABCB10
Gene Name
ATP binding cassette subfamily B member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012089.2 Intron NP_036221.2
Gene
LOC101927478
Gene Name
uncharacterized LOC101927478
There are no transcripts associated with this gene.

Gene
NUP133
Gene Name
nucleoporin 133
There are no transcripts associated with this gene.

View Full Product Details