Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142473.1 | 1347 | UTR 3 | NP_001135945.1 | ||
NM_001193338.1 | 1347 | UTR 3 | NP_001180267.1 | ||
NM_005449.4 | 1347 | UTR 3 | NP_005440.1 | ||
XM_005273351.3 | 1347 | UTR 3 | XP_005273408.1 | ||
XM_005273352.4 | 1347 | Intron | XP_005273409.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001185156.1 | 1347 | UTR 3 | NP_001172085.1 | ||
NM_001185157.1 | 1347 | UTR 3 | NP_001172086.1 | ||
NM_001185158.1 | 1347 | UTR 3 | NP_001172087.1 | ||
NM_006850.3 | 1347 | UTR 3 | NP_006841.1 | ||
XM_011509101.2 | 1347 | Intron | XP_011507403.1 | ||
XM_017000121.1 | 1347 | Intron | XP_016855610.1 |