Product Details

SNP ID
rs4565707
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:157795442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGAATAAAAGAAGACACACTGT[A/T]TTTATAAAGCTTCTAGGGGCTGTAA
Phenotype
MIM: 606508
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FCRL1 PubMed Links

Gene Details

Gene
FCRL1
Gene Name
Fc receptor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159397.1 2057 UTR 3 NP_001152869.1
NM_001159398.1 2057 UTR 3 NP_001152870.1
NM_052938.4 2057 UTR 3 NP_443170.1

View Full Product Details