Product Details

SNP ID
rs3829642
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:10461342 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGGGCTAAAAAAAAAATTGGTGGAA[C/T]GGCGTATGTTGAGACCTGGAATAGC
Phenotype
MIM: 602784 MIM: 601882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APITD1-CORT PubMed Links

Gene Details

Gene
APITD1-CORT
Gene Name
APITD1-CORT readthrough
There are no transcripts associated with this gene.

Gene
CORT
Gene Name
cortistatin
There are no transcripts associated with this gene.

Gene
DFFA
Gene Name
DNA fragmentation factor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004401.2 1242 UTR 3 NP_004392.1
NM_213566.1 1242 UTR 3 NP_998731.1

View Full Product Details