Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001277081.1 | 1857 | UTR 3 | NP_001264010.1 | ||
NM_001277082.1 | 1857 | UTR 3 | NP_001264011.1 | ||
NM_017509.3 | 1857 | UTR 3 | NP_059979.2 | ||
XM_006723265.3 | 1857 | UTR 3 | XP_006723328.1 | ||
XM_011527085.2 | 1857 | UTR 3 | XP_011525387.1 | ||
XM_011527087.2 | 1857 | UTR 3 | XP_011525389.1 | ||
XM_011527088.2 | 1857 | Intron | XP_011525390.1 | ||
XM_011527089.2 | 1857 | UTR 3 | XP_011525391.1 | ||
XM_017026943.1 | 1857 | Intron | XP_016882432.1 |