Product Details

SNP ID
rs3212847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50817741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCTTTTTTTGTCCCCTCTCTCC[A/G]GGTATCTTTCTGTCTGTTGTGAGGG
Phenotype
MIM: 147910 MIM: 610601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK1 PubMed Links

Gene Details

Gene
KLK1
Gene Name
kallikrein 1
There are no transcripts associated with this gene.

Gene
KLK15
Gene Name
kallikrein related peptidase 15
There are no transcripts associated with this gene.

Gene
MGC45922
Gene Name
uncharacterized LOC284365
There are no transcripts associated with this gene.

View Full Product Details