Product Details

SNP ID
rs3093724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28347605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAAGAGTCCAGTCCAGCACCCC[C/T]GCCAACTAGCTGAGAGGCACAGTGA
Phenotype
MIM: 611519 MIM: 616815 MIM: 191161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLDIP2 PubMed Links

Gene Details

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290145.1 1706 UTR 3 NP_001277074.1
NM_015584.4 1706 UTR 3 NP_056399.1
Gene
TMEM199
Gene Name
transmembrane protein 199
There are no transcripts associated with this gene.

Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021137.4 1706 Intron NP_066960.1
XM_017024993.1 1706 Intron XP_016880482.1

View Full Product Details