Product Details

SNP ID
rs3809796
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29255917 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTACAAATTTCTCAAGTTTTTTTT[C/T]TGATAAAATAAGTAAATCTGGGTAT
Phenotype
MIM: 123610 MIM: 609356
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CRYBA1 PubMed Links

Gene Details

Gene
CRYBA1
Gene Name
crystallin beta A1
There are no transcripts associated with this gene.

Gene
NUFIP2
Gene Name
NUFIP2, FMR1 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020772.2 12097 UTR 3 NP_065823.1
XM_017024896.1 12097 UTR 3 XP_016880385.1

View Full Product Details