Product Details
- SNP ID
-
rs4970925
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:150493798 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAAATACTGTTCTGGACAAGAAGA[C/G]TTACTAGATGGAATGGGATGAGTTA
- Phenotype
-
MIM: 612805
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6878
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114187507,rs115154028] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6878
- Gene Name
- microRNA 6878
There are no transcripts associated with this gene.
- Gene
- TARS2
- Gene Name
- threonyl-tRNA synthetase 2, mitochondrial (putative)
View Full Product Details