Product Details

SNP ID
rs3177563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50586298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAACCCTAGGTTCTCTCTGCAC[C/T]ATTAACACAGACATCTCAGGACATG
Phenotype
MIM: 176885
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM65C PubMed Links

Gene Details

Gene
FAM65C
Gene Name
family with sequence similarity 65 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290268.1 4189 UTR 3 NP_001277197.1
NM_080829.3 4189 UTR 3 NP_543019.2
XM_005260294.3 4189 UTR 3 XP_005260351.1
XM_006723713.3 4189 UTR 3 XP_006723776.1
XM_011528578.2 4189 UTR 3 XP_011526880.1
XM_011528579.2 4189 UTR 3 XP_011526881.1
XM_011528580.2 4189 UTR 3 XP_011526882.1
XM_011528581.2 4189 UTR 3 XP_011526883.1
XM_011528584.2 4189 Intron XP_011526886.1
XM_011528585.2 4189 UTR 3 XP_011526887.1
XM_011528586.2 4189 Intron XP_011526888.1
XM_017027681.1 4189 UTR 3 XP_016883170.1
XM_017027682.1 4189 Intron XP_016883171.1
Gene
MIR645
Gene Name
microRNA 645
There are no transcripts associated with this gene.

Gene
PTPN1
Gene Name
protein tyrosine phosphatase, non-receptor type 1
There are no transcripts associated with this gene.

View Full Product Details