Product Details

SNP ID
rs3209551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50192588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAATCTATTTCTATGAGAAAA[A/G]AGGCGTCTGTATATTTTGGGAATCT
Phenotype
MIM: 189965
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEBPB PubMed Links

Gene Details

Gene
CEBPB
Gene Name
CCAAT/enhancer binding protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285878.1 2006 UTR 3 NP_001272807.1
NM_001285879.1 2006 UTR 3 NP_001272808.1
NM_005194.3 2006 UTR 3 NP_005185.2
Gene
CEBPB-AS1
Gene Name
CEBPB antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details