Product Details

SNP ID
rs4253440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50191616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAGGAGGCCGGGGCGCCGGGC[A/G]GCGGCGCAGGCATGGCGGCGGGCTT
Phenotype
MIM: 189965
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEBPB PubMed Links

Gene Details

Gene
CEBPB
Gene Name
CCAAT/enhancer binding protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285878.1 1034 Missense Mutation AGC,GGC S172G NP_001272807.1
NM_001285879.1 1034 UTR 5 NP_001272808.1
NM_005194.3 1034 Missense Mutation AGC,GGC S195G NP_005185.2
Gene
CEBPB-AS1
Gene Name
CEBPB antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details