Product Details
- SNP ID
-
rs4784146
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:61648991 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTCTTCTACAATTATCATAAGAA[A/G]CAATAATTACAAGGGCAAATAATAC
- Phenotype
-
MIM: 603008
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH8
PubMed Links
Gene Details
- Gene
- CDH8
- Gene Name
- cadherin 8
View Full Product Details