Product Details

SNP ID
rs3801791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:111727417 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTGTCTGTATTGATTCCTGAATT[C/G]TTGCTAATGGGGCACAGTCATTTGT
Phenotype
MIM: 607679
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DOCK4 PubMed Links

Gene Details

Gene
DOCK4
Gene Name
dedicator of cytokinesis 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014705.3 7559 UTR 3 NP_055520.3
XM_006716188.2 7559 UTR 3 XP_006716251.1
XM_006716189.2 7559 UTR 3 XP_006716252.1
XM_017012819.1 7559 UTR 3 XP_016868308.1
XM_017012820.1 7559 UTR 3 XP_016868309.1
XM_017012821.1 7559 UTR 3 XP_016868310.1
XM_017012822.1 7559 UTR 3 XP_016868311.1
XM_017012823.1 7559 UTR 3 XP_016868312.1
XM_017012824.1 7559 UTR 3 XP_016868313.1
XM_017012825.1 7559 UTR 3 XP_016868314.1

View Full Product Details