Product Details

SNP ID
rs7803697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140700870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAATGTAATATCTATATGCACAG[C/G]TAGATCCTGAATGAATAATATAATT
Phenotype
MIM: 603838
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADCK2 PubMed Links

Gene Details

Gene
ADCK2
Gene Name
aarF domain containing kinase 2
There are no transcripts associated with this gene.

Gene
NDUFB2
Gene Name
NADH:ubiquinone oxidoreductase subunit B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004546.2 Intron NP_004537.1
Gene
NDUFB2-AS1
Gene Name
NDUFB2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details