Product Details
- SNP ID
-
hCV32355808
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:133255858 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACGGCCTCGATGCCGTTGGCCTG[G/C]TCGACCATCATGGCCTGGTGGCAGG
- Phenotype
-
MIM: 110300
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8176745] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABO
- Gene Name
- ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020469.2 |
898 |
Missense Mutation |
GAC,GAG |
D291E |
NP_065202.2 |
View Full Product Details