Product Details

SNP ID
rs4838930
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:110158099 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTTGTCTGATTTCTATTTGTCC[C/G]TCTGCACTAGAAATAAATCCAATGG
Phenotype
MIM: 610299
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC6A17 PubMed Links

Gene Details

Gene
SLC6A17
Gene Name
solute carrier family 6 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010898.2 Intron NP_001010898.1
XM_006710643.2 Intron XP_006710706.1

View Full Product Details