Product Details

SNP ID
rs3730683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54271000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTTAGAATAATTTTCAAGCTGT[A/G]TGAAAATCAACGTCATTTCAACTTT
Phenotype
MIM: 605252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLI PubMed Links

Gene Details

Gene
POLI
Gene Name
DNA polymerase iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007195.2 Intron NP_009126.2
XM_005258188.3 Intron XP_005258245.1
XM_005258192.4 Intron XP_005258249.1
XM_006722378.2 Intron XP_006722441.1
XM_011525796.2 Intron XP_011524098.1
XM_011525797.1 Intron XP_011524099.1

View Full Product Details