Product Details

SNP ID
hCV32368187
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88708712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATCCATTCAACGTAGGGAGGGA[C/T]TGCTGTGCACATCAAGTGTGTCTGA
Phenotype
MIM: 611184
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CTU2 PubMed Links

Gene Details

Gene
CTU2
Gene Name
cytosolic thiouridylase subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012759.2 Intron NP_001012777.1
NM_001012762.2 Intron NP_001012780.1
NM_001318507.1 Intron NP_001305436.1
NM_001318513.1 Intron NP_001305442.1
Gene
MIR4722
Gene Name
microRNA 4722
There are no transcripts associated with this gene.

Gene
PIEZO1
Gene Name
piezo type mechanosensitive ion channel component 1
There are no transcripts associated with this gene.

Gene
RNF166
Gene Name
ring finger protein 166
There are no transcripts associated with this gene.

View Full Product Details