Product Details

SNP ID
rs7946766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47982817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTGTATTTCATTAGCATATTGC[C/T]ACTCTACACTTGTCCTGTATTTAGA
Phenotype
MIM: 600925
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRJ PubMed Links

Gene Details

Gene
PTPRJ
Gene Name
protein tyrosine phosphatase, receptor type J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098503.1 Intron NP_001091973.1
NM_002843.3 Intron NP_002834.3
XM_017018083.1 Intron XP_016873572.1
XM_017018084.1 Intron XP_016873573.1
XM_017018085.1 Intron XP_016873574.1

View Full Product Details