Product Details

SNP ID
rs4149856
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:241849687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTTAATTGAAATACTTACCTT[C/T]CTACAAACTGTTTTGAGTTTCTACT
Phenotype
MIM: 606063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXO1 PubMed Links

Gene Details

Gene
EXO1
Gene Name
exonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319224.1 Intron NP_001306153.1
NM_003686.4 Intron NP_003677.4
NM_006027.4 Intron NP_006018.4
NM_130398.3 Intron NP_569082.2
XM_006711840.2 Intron XP_006711903.1
XM_011544321.1 Intron XP_011542623.1
XM_011544322.1 Intron XP_011542624.1
XM_011544323.2 Intron XP_011542625.1
XM_011544324.2 Intron XP_011542626.1
XM_011544325.2 Intron XP_011542627.1
XM_017002793.1 Intron XP_016858282.1

View Full Product Details