Product Details

SNP ID
rs4328567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54237548 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAAGTGGATTGAGGGTCACCAAAT[C/G]AAAGTCAGCGTTCCAGTGCCCGCGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LILRA6 PubMed Links

Gene Details

Gene
LILRA6
Gene Name
leukocyte immunoglobulin like receptor A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024318.3 Intron NP_077294.3

View Full Product Details