Product Details

SNP ID
rs4683639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141880663 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTTTACCTAATAGGTTCATTC[C/T]AGCTTTAAAAAAAGTGTTGTTTCAT
Phenotype
MIM: 601867
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP1B3 PubMed Links

Gene Details

Gene
ATP1B3
Gene Name
ATPase Na+/K+ transporting subunit beta 3
There are no transcripts associated with this gene.

View Full Product Details