Product Details

SNP ID
rs3730397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67194225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTTCCTGGTTACTGACCCTGGG[C/T]TCTCAGGAAGAGAAGAGCTTTAGCT
Phenotype
MIM: 600659 MIM: 606422 MIM: 614117 MIM: 613701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
E2F4 PubMed Links

Gene Details

Gene
E2F4
Gene Name
E2F transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001950.3 Intron NP_001941.2
Gene
ELMO3
Gene Name
engulfment and cell motility 3
There are no transcripts associated with this gene.

Gene
EXOC3L1
Gene Name
exocyst complex component 3 like 1
There are no transcripts associated with this gene.

Gene
MIR328
Gene Name
microRNA 328
There are no transcripts associated with this gene.

View Full Product Details