Product Details

SNP ID
rs3218214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10567974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGACTTCCCTGTAAAATGTCTCC[C/T]AGGGAATATAGGGATGTCTTGCAAT
Phenotype
MIM: 607309 MIM: 600927
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP1M2 PubMed Links

Gene Details

Gene
AP1M2
Gene Name
adaptor related protein complex 1 mu 2 subunit
There are no transcripts associated with this gene.

Gene
CDKN2D
Gene Name
cyclin dependent kinase inhibitor 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001800.3 Intron NP_001791.1
NM_079421.2 Intron NP_524145.1
Gene
KRI1
Gene Name
KRI1 homolog
There are no transcripts associated with this gene.

View Full Product Details