Product Details
- SNP ID
-
rs3181012
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:16325384 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATTACCCCGAACCCGGCGCGCCC[A/C]CGCCCTACAGCGCCCCCGCGGGTGG
- Phenotype
-
MIM: 602016
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLF2
PubMed Links
Gene Details
- Gene
- KLF2
- Gene Name
- Kruppel like factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016270.3 |
342 |
Missense Mutation |
ACG,CCG |
T82P |
NP_057354.1 |
View Full Product Details