Product Details
- SNP ID
-
hCV32378028
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:121843897 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCAACTCCCCTAGCCAGGTGGA[G/T]GTGCTGGGTCATCAGGATACAGGGT
- Phenotype
-
MIM: 609695
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HPD
PubMed Links
Gene Details
- Gene
- HPD
- Gene Name
- 4-hydroxyphenylpyruvate dioxygenase
View Full Product Details