Product Details

SNP ID
rs3755026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38069805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGATAATACTAATATACTGCTTA[C/T]GGACAGTTAAGTAATATATTTCACA
Phenotype
MIM: 601771 MIM: 611872
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP1B1 PubMed Links

Gene Details

Gene
CYP1B1
Gene Name
cytochrome P450 family 1 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000104.3 2952 UTR 3 NP_000095.2
Gene
RMDN2
Gene Name
regulator of microtubule dynamics 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170791.2 2952 Intron NP_001164262.1
NM_001170792.2 2952 Intron NP_001164263.1
NM_001170793.2 2952 Intron NP_001164264.1
NM_001322211.1 2952 Intron NP_001309140.1
NM_001322212.1 2952 Intron NP_001309141.1
NM_144713.4 2952 Intron NP_653314.3

View Full Product Details