Product Details

SNP ID
rs4252215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69370295 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTTTTAAAGGAGGTAATTGTCA[A/G]TCTTGTTTTCAAGAGTGAATAGAGA
Phenotype
MIM: 603139 MIM: 600822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AK6 PubMed Links

Gene Details

Gene
AK6
Gene Name
adenylate kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015891.1 Intron NP_001015891.1
NM_016283.4 Intron NP_057367.1
Gene
RAD17
Gene Name
RAD17 checkpoint clamp loader component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278622.1 Intron NP_001265551.1
NM_002873.1 Intron NP_002864.1
NM_133338.2 Intron NP_579916.1
NM_133339.2 Intron NP_579917.1
NM_133340.2 Intron NP_579918.1
NM_133341.2 Intron NP_579919.1
NM_133342.2 Intron NP_579920.1
NM_133343.1 Intron NP_579921.1
NM_133344.2 Intron NP_579922.1
XM_017009679.1 Intron XP_016865168.1
XM_017009680.1 Intron XP_016865169.1
XM_017009681.1 Intron XP_016865170.1
Gene
TAF9
Gene Name
TATA-box binding protein associated factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015892.1 Intron NP_001015892.1
NM_003187.4 Intron NP_003178.1

View Full Product Details