Product Details

SNP ID
rs4752268
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114828978 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGACCAATATTTCTAATTTGGACA[A/G]TAGTTCGGAAATTGTTTCGACAGGG
Phenotype
MIM: 602330
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ABLIM1 PubMed Links

Gene Details

Gene
ABLIM1
Gene Name
actin binding LIM protein 1
There are no transcripts associated with this gene.

Gene
FAM160B1
Gene Name
family with sequence similarity 160 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135051.1 Intron NP_001128523.1
NM_020940.3 Intron NP_065991.3

View Full Product Details