Product Details
- SNP ID
-
rs4272763
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:34441390 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCATTCAAGTTGTTTCATGTAT[A/T]AGCAGTTTGTTCTTTTCATTGCTGA
- Phenotype
-
MIM: 115500
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CAT
PubMed Links
Gene Details
- Gene
- CAT
- Gene Name
- catalase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001752.3 |
|
Intron |
|
|
NP_001743.1 |
View Full Product Details