Product Details
- SNP ID
-
rs140072597
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:92213315 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGTCCTGTGAAAAATGCTGCTCC[A/G]TAAGTAGGAATTTCCCAGCAATTCT
- Phenotype
-
MIM: 604214
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRIT1
PubMed Links
Gene Details
- Gene
- KRIT1
- Gene Name
- KRIT1, ankyrin repeat containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001013406.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y587Y |
NP_001013424.1 |
NM_004912.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
NP_004903.2 |
NM_194454.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
NP_919436.1 |
NM_194455.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
NP_919437.1 |
NM_194456.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
NP_919438.1 |
XM_005250660.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_005250717.1 |
XM_005250662.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_005250719.1 |
XM_005250665.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_005250722.1 |
XM_005250666.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_005250723.1 |
XM_005250667.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_005250724.1 |
XM_005250668.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_005250725.1 |
XM_005250669.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_005250726.1 |
XM_006716161.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_006716224.1 |
XM_006716162.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_006716225.1 |
XM_006716163.3 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_006716226.1 |
XM_011516651.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514953.1 |
XM_011516653.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514955.1 |
XM_011516654.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514956.1 |
XM_011516655.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514957.1 |
XM_011516656.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514958.1 |
XM_011516657.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514959.1 |
XM_011516658.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514960.1 |
XM_011516659.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514961.1 |
XM_011516660.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514962.1 |
XM_011516661.2 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_011514963.1 |
XM_017012755.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_016868244.1 |
XM_017012756.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_016868245.1 |
XM_017012757.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y635Y |
XP_016868246.1 |
XM_017012758.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y397Y |
XP_016868247.1 |
XM_017012759.1 |
2644 |
Silent Mutation |
TAC,TAT |
Y397Y |
XP_016868248.1 |
- Gene
- MIR1285-1
- Gene Name
- microRNA 1285-1
There are no transcripts associated with this gene.
View Full Product Details