Product Details

SNP ID
rs4904783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91457951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAAAACTCTAAACAAATTAAGA[A/C]CTGTGATGTAGAACAAAAATTACAC
Phenotype
MIM: 610351
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPP4R3A PubMed Links

Gene Details

Gene
PPP4R3A
Gene Name
protein phosphatase 4 regulatory subunit 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284280.1 3426 UTR 3 NP_001271209.1
NM_001284281.1 3426 UTR 3 NP_001271210.1

View Full Product Details