Product Details

SNP ID
rs4255898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19887793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTTTTTTTTTTTTTTTTTTTA[A/G]AAGAAGTCTCACTCTGCCACTCAGA
Phenotype
MIM: 606954
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF253 PubMed Links

Gene Details

Gene
ZNF253
Gene Name
zinc finger protein 253
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021047.2 Intron NP_066385.2

View Full Product Details