Product Details

SNP ID
rs4150978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:85215468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGTCAGATAAAAACACAGGGTTA[A/G]AGGATTAGGACTTTCTGCTCCAGAG
Phenotype
MIM: 600967
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8orf59 PubMed Links

Gene Details

Gene
C8orf59
Gene Name
chromosome 8 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099670.2 Intron NP_001093140.1
NM_001099671.2 Intron NP_001093141.1
NM_001099672.2 Intron NP_001093142.1
NM_001099673.2 Intron NP_001093143.1
NM_001293320.1 Intron NP_001280249.1
XM_011517528.2 Intron XP_011515830.1
Gene
E2F5
Gene Name
E2F transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083588.1 Intron NP_001077057.1
NM_001083589.1 Intron NP_001077058.1
NM_001951.3 Intron NP_001942.2

View Full Product Details