Product Details

SNP ID
rs1805747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8996321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTGCCTAGTATAAAAATCAATC[A/G]AATTCTAATTTGTCAACATAAATAA
Phenotype
MIM: 604874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRG1 PubMed Links

Gene Details

Gene
KLRG1
Gene Name
killer cell lectin like receptor G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005810.3 Intron NP_005801.3
XM_017018682.1 Intron XP_016874171.1
XM_017018683.1 Intron XP_016874172.1
XM_017018684.1 Intron XP_016874173.1
XM_017018685.1 Intron XP_016874174.1

View Full Product Details