Product Details

SNP ID
rs3767336
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:165403728 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CAAAGGAAACAGCTTCTCTGTAAGT[A/G]CAAAGTTAGACTGGGATTAACTTTA
Phenotype
MIM: 180247
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RXRG PubMed Links

Gene Details

Gene
RXRG
Gene Name
retinoid X receptor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256570.1 Intron NP_001243499.1
NM_001256571.1 Intron NP_001243500.1
NM_006917.4 Intron NP_008848.1

View Full Product Details