Product Details
- SNP ID
-
rs3917431
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:169726240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGTGAGATTTCCTATTTTGTAAA[C/T]GTCAGAACTTAACTCAAAAATGTTT
- Phenotype
-
MIM: 131210
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SELE
PubMed Links
Gene Details
- Gene
- SELE
- Gene Name
- selectin E
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000450.2 |
|
Intron |
|
|
NP_000441.2 |
View Full Product Details