Product Details
- SNP ID
-
hCV32399511
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:54072400 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAAAGGCCATTGATAGTTTTTTT[A/G]AAGTGTATAAAATAAAAAATAAAAA
- Phenotype
-
MIM: 300741
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM120C
PubMed Links
Gene Details
- Gene
- FAM120C
- Gene Name
- family with sequence similarity 120C
There are no transcripts associated with this gene.
View Full Product Details