Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098268.1 | 3525 | UTR 3 | NP_001091738.1 | ||
NM_002312.3 | 3525 | UTR 3 | NP_002303.2 | ||
NM_206937.1 | 3525 | UTR 3 | NP_996820.1 | ||
XM_005254056.1 | 3525 | UTR 3 | XP_005254113.1 | ||
XM_005254057.4 | 3525 | UTR 3 | XP_005254114.1 | ||
XM_005254058.3 | 3525 | UTR 3 | XP_005254115.1 | ||
XM_006719951.3 | 3525 | UTR 3 | XP_006720014.1 | ||
XM_006719952.1 | 3525 | UTR 3 | XP_006720015.1 | ||
XM_011521091.2 | 3525 | UTR 3 | XP_011519393.1 | ||
XM_011521092.2 | 3525 | UTR 3 | XP_011519394.1 | ||
XM_017020563.1 | 3525 | UTR 3 | XP_016876052.1 | ||
XM_017020564.1 | 3525 | UTR 3 | XP_016876053.1 | ||
XM_017020565.1 | 3525 | UTR 3 | XP_016876054.1 | ||
XM_017020566.1 | 3525 | UTR 3 | XP_016876055.1 | ||
XM_017020567.1 | 3525 | UTR 3 | XP_016876056.1 | ||
XM_017020568.1 | 3525 | UTR 3 | XP_016876057.1 | ||
XM_017020569.1 | 3525 | UTR 3 | XP_016876058.1 | ||
XM_017020570.1 | 3525 | UTR 3 | XP_016876059.1 | ||
XM_017020571.1 | 3525 | UTR 3 | XP_016876060.1 | ||
XM_017020572.1 | 3525 | UTR 3 | XP_016876061.1 | ||
XM_017020573.1 | 3525 | UTR 3 | XP_016876062.1 |